missing translation for 'onlineSavingsMsg'
Learn More

Silicon Standard For ICP, 1000μg/mL in H2O/0.24 F, Assurance Grade, SPEX CertiPrep™

Product Code. 10325792 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
125 mL
500 mL
Unit Size:
125mL
500mL
This item is not returnable. View return policy

Product Code. 10325792

Brand: SPEX Certiprep PLSI92X

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Si1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Silicon Standard
Packaging Triple-leached LDPE bottle
CAS 16919-19-0, 7732-18-5
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
CAS Min % 0.1, 99.9
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 0.1, 99.9
Molecular Formula F6H8N2Si
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key ITHIMUMYFVCXSL-UHFFFAOYSA-P
Type Certified Reference Material
SMILES [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
IUPAC Name diazanium;hexafluorosilicon(2-)
Solvent or Matrix H2O/0.4% F
Molecular Weight (g/mol) 178.153
PubChem CID 28145
Quantity 500 mL
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.H2O/tr. HF    1000μg/mL    PLSI9-2Y, -2T, -2X    10,000    PLSI9-3Y, -3XH2O    1000μg/mL    PLSI9A-2X    10,000    PLSI9A-3X
Show More Show Less

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.