missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Door Gasket

Material: FKM Fluorocarbon; For Use With: FD 23

Brand:  BINDER™ 8012-1306

395.20 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.



Additional Details : Weight : 7.50000kg

Product Code. 16464169

  • £416.00 / Each
Estimated Shipment: 16-10-2024
to see stock.
Add to basket

Description

Description

  • Manufactured from FKM fluorocarbon
  • Temperature-resistant up to 200°C
Specifications

Specifications

Door gasket - made of FKM, silicone-free
BINDER Chambers FD 23
FKM Fluorocarbon
Door Gasket
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers