Learn More
BINDER™ Door Gasket
Material: FKM Fluorocarbon; For Use With: M 240
Brand:  BINDER™ 8012-1309
Product Code. 16494169
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Manufactured from FKM fluorocarbon
  • Temperature-resistant up to 200°C
Specifications
Specifications
Door gasket - made of FKM, silicone-free
BINDER Chambers M 240
FKM Fluorocarbon
Door Gasket
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Ofertas especiales
Ofertas especiales