Learn More
Thermo Scientific Chemicals Silicone Grease, medium viscosity, Tube with 35gr
Brand: Thermo Scientific Chemicals 386110010
19.65 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Code : 29
Additional Details : Weight : 0.05264kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silicone Grease | |
1 Ea. | |
1.23 oz. |
RUO – Research Use Only