Learn More
Macherey-Nagel™ Silicone High Temperature Septa
Brand:  Macherey-Nagel™ 702623
Product Code. 10206501
  • £67.70 / Pack of 50
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
High Temperature Septum
60° Shore A
Red
50 Pack
13 mm
320°C
Silicone