Learn More
Silicone oil, for oil baths, usable range from -40 to +200°C, Thermo Scientific Chemicals
For oil baths
£22.80 - £172.00
Chemical Identifiers
CAS | 63148-62-9 |
---|---|
Molecular Formula | (C2H6OSi)n |
Molecular Weight (g/mol) | NaN |
MDL Number | MFCD00132673 |
IUPAC Name | Polydimethylsiloxane |
SMILES | C[Si](C)(-*)O-* |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11460673
|
Thermo Scientific Alfa Aesar
A12728.22 |
100 g |
£22.80
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470673
|
Thermo Scientific Alfa Aesar
A12728.36 |
500 g |
£55.50
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480673
|
Thermo Scientific Alfa Aesar
A12728.0E |
2500 g |
£172.00
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Silicone oil is used as a calibration media in temperature controlled baths in laboratories due to its good thermal characteristics and low flammability. It is also used as active component in silicone grease, lubricants and heat transfer fluids. It finds application in cosmetics, hair conditioners and as a sorbent for the analysis of headspace (dissolved gas analysis) of food.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSilicone oil is used as a calibration media in temperature controlled baths in laboratories due to its good thermal characteristics and low flammability. It is also used as active component in silicone grease, lubricants and heat transfer fluids. It finds application in cosmetics, hair conditioners and as a sorbent for the analysis of headspace (dissolved gas analysis) of food.
Solubility
Miscible with aliphatic, aromatic hydrocarbons, alcohol and chlorinated hydrocarbons. Immiscible with water, methanol and ethylene glycol.
Notes
Viscosity at 40cSt. Incompatible with strong oxidizing agents.
Chemical Identifiers
63148-62-9 | |
NaN | |
Polydimethylsiloxane |
(C2H6OSi)n | |
MFCD00132673 | |
C[Si](C)(-*)O-* |
Specifications
63148-62-9 | |
>270°C (518°F) | |
1.404 | |
Miscible with aliphatic,aromatic hydrocarbons,alcohol and chlorinated hydrocarbons. Immiscible with water,methanol and ethylene glycol. | |
Polydimethylsiloxane | |
Silicone Oil |
0.963 | |
(C2H6OSi)n | |
MFCD00132673 | |
C[Si](C)(-*)O-* | |
NaN |
Safety and Handling
missing translation for 'rtecsNumber' : JT6484500
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only