missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ismatec™ Peroxide-Cured Silicone Pump Tubing
Reduce the risk of particulate entrapment with smooth inner surface
Brand: Ismatec™ 07625-34
Code : XX
Additional Details : Weight : 0.01000kg
Description
- Tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Regulatory approvals to match your unique application
Specifications
Yes | |
Silicone | |
Masterflex Silicone (peroxide cured) color-coded stopped tubing for Ismatec pumps | |
34 |
1.14 mm | |
15 m | |
extending the length of tubing |