missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Replacement Pipette Adapter

A replacement pipette adapter for use with the RF1000 Pipette Controller.

Brand:  Fisherbrand™ HS5000RFRP

Product Code. 11831920

  • £20.45 / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Manufactured from silicone rubber
  • Aids in manual one-handed pipetting
  • Allows for a large range of pipette sizes
Specifications

Specifications

1 to 50mL glass and plastic pipettes can be used
Silicone Rubber
1 Each
Single-use
1 to 50 mL
Allows for a large range of pipette sizes
Adapter
RF1000 Manual Pipette Controller
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Replacement Pipette Adapter > For Use With (Equipment): For Use with RF1000 Pipette Controller

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.