missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ HotGrip™ Hand Protectors

Catalog Number p-8002758
36.15 GBP valid until 2024-12-31
Use promo code "23267" to get your promotional price.


Click to view available options
Color:
Red
White
For Use With (Application):
Protects The Whole Hand From Burns and Freezing
Protects Thumb and 1 Finger From Burns and Freezing
Protects Thumb and 2 to 3 Fingers From Burns and Freezing
This item is not returnable. View return policy
This item is not returnable. View return policy

Manufactured from silicone rubber for extreme thermal resistance. Buerkle™ HotGrip™ Hand Protectors protect fingers and hands from burns and freezing.

  • Resistant to temperatures from –50 to +200°C (briefly up to +250°C)
  • Deep frozen, hot or boiling substances and apparatus can be safely handled and positioned
  • Sililcone rubber is extremely elastic and slip-proof; ideal for smooth surfaces
  • Secure grip 
Material Silicone Rubber
Color Red
For Use With (Application) Protects The Whole Hand From Burns and Freezing
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'