Learn More
Cole Parmer Silicone Sanitary Gasket, Tri-Clamp
Cole Parmer™ Silicone Sanitary Gasket is designed for Tri-Clamp™ style fittings and comes in a wide variety of materials to match specific process needs.
£36.10
Specifications
Certifications/Compliance | FDA |
---|---|
Material | Silicone |
Quantity | 10/Pk. |
Sterility | Non-sterile |
Type | Gasket |
Description
Cole Parmer™ Silicone Sanitary Gasket is designed for Tri-Clamp™ style fittings and comes in a wide variety of materials to match specific process needs.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Single-use custom configuration solutions to minimize downtime
- All gaskets meet USDA, 3-A and CGMP criteria
- FDA Compliant
- All silicone, EPDM, PTFE, and Viton™ gaskets are animal-derived ingredient free and meet USP Class IV.
Specifications
FDA | |
10/Pk. | |
Gasket |
Silicone | |
Non-sterile |