Learn More
DWK Life Sciences Silicone Sealing Ring, VMQ
The silicone seal is used for sealing and fixing threaded pipes with threaded caps.
Brand:  DWK Life Sciences 292361407
 View more versions of this product
Product Code. 12851054
  • £94.80 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
The silicone sealing ring matches the screw connection piece and has good sealing properties. Due to the PTFE layer on one side of the sealing ring, there is a certain inertness of the material. The included hole allows the insertion of glass rods, olives, hoses, etc.
  • With bonded PTFE face
TRUSTED_SUSTAINABILITY
Specifications
Specifications
13.0 to 15.0 mm
GL 32
Fits Tubing ID: 13.0 to 15.0 mm
Silicone (VMQ)
Sealing; Connecting glass tubes; Suitable for PBT screw cap with aperture
10/Pk.
29 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences Silicone Sealing Ring, VMQ > GL 32, fits tube ID 13.0 to 15.0 mm