missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Silicone Sealing Ring, VMQ

The silicone seal is used for sealing and fixing threaded pipes with threaded caps.

Brand:  DWK Life Sciences 292361604

 View more versions of this product

Product Code. 13157444

  • £88.30 / Pack of 10
Estimated Shipment: 26-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The silicone sealing ring matches the screw connection piece and has good sealing properties. Due to the PTFE layer on one side of the sealing ring, there is a certain inertness of the material. The included hole allows the insertion of glass rods, olives, hoses, etc.

  • With bonded PTFE face
TRUSTED_SUSTAINABILITY
Specifications

Specifications

15.0 to 17.0 mm
GL 32
Fits Tubing ID: 15.0 to 17.0 mm
Silicone (VMQ)
Sealing; Connecting glass tubes; Suitable for PBT screw cap with aperture
10/Pk.
29 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Silicone Sealing Ring, VMQ > GL 32, fits tube ID 15.0 to 17.0 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.