missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Silicone Stoppers

Product Code. 10757311 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Bottom:
3 mm
6 mm
8 mm
13 mm
17 mm
31 mm
43 mm
49 mm
53 mm
Quantity:
1/Pk.
10/Pk.
2/Pk.
20/Pk.
50/Pk.
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 20
Pack of 50

Product Code. 10757311

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

For laboratory and/or food testing, industrial masking or educational use.

  • Non-toxic, translucent
  • Temperature range -60°C to 200°C
  • High electrical resistance
  • Resists cracking and spallation
  • High resistance to many chemicals including ozone

Specifications

Quantity 50/Pk.
Diameter (Metric) Bottom 6 mm
Material Silicone
Product Type Stopper
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.