Learn More
Hirschmann™ SILICOSEN™ Pink Silicone Stoppers
Excellent for the cultivation of aerobic microorganisms. Hirschmann Instruments™ SILICOSEN™ Silicone Stoppers are specially processed culture plugs with continuous bubbles for preparation, filling and sterilization of culture media.
Brand:  Hirschmann™ 8905617
 View more versions of this product
Product Code. 10238730
  • £53.97 / Pack of 10
Explore more special offers
Description
Description
  • Uniform pore structure
  • Good air permeability
  • Chemically and temperature resistant
  • Liquid repellent and reusable
Specifications
Specifications
13-16 mm
30 mm
Pink
Silicone Stopper
For The Preparation And Sterilization Of Culture Media
10/Pk.
Silicone