Learn More
Hirschmann™ SILICOSEN™ Pink Silicone Stoppers
Excellent for the cultivation of aerobic microorganisms. Hirschmann Instruments™ SILICOSEN™ Silicone Stoppers are specially processed culture plugs with continuous bubbles for preparation, filling and sterilization of culture media.
Brand: Hirschmann™ 8905524
This item is not returnable.
View return policy
Description
- Uniform pore structure
- Good air permeability
- Chemically and temperature resistant
- Liquid repellent and reusable
Specifications
20-23 mm | |
20 mm | |
Pink | |
Silicone Stopper |
For The Preparation And Sterilization Of Culture Media | |
10/Pk. | |
Silicone |