Learn More
Masterflex™ Silicone Tubing
Use this tubing for pharmaceutical, biotechnology, food and beverage, aerospace and general lab applications. Saint Gobain Performance Plastics™ Silicone Tubing is peroxide-cured, offers excellent heat resistance and a low compression set.
Brand:  Masterflex™ 06411-60
 View more versions of this product
Product Code. 11751179
  • £221.00 / 7.50m
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use this tubing for pharmaceutical, biotechnology, food and beverage, aerospace and general lab applications. Saint Gobain Performance Plastics™ Silicone Tubing is peroxide-cured, offers excellent heat resistance and a low compression set.
  • Odorless, nontoxic, translucent tubing
  • Elastomer is FDA-compliant (21 CFR 177.2600) and meets USP Class VI requirements
  • Manufactured according to GMP
  • Sterilize by autoclave or gamma irradiation
Specifications
Specifications
Autoclavable
Silicone
7.6 m
-50°C
230°c
0.8 mm
4 mm
0.8 mm
Translucent