missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Silicone Tubing

Use this tubing for pharmaceutical, biotechnology, food and beverage, aerospace and general lab applications. Saint Gobain Performance Plastics™ Silicone Tubing is peroxide-cured, offers excellent heat resistance and a low compression set.

Brand:  Masterflex™ 06411-03

 View more versions of this product

Product Code. 11755948

  • £183.00 / 7.50m
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use this tubing for pharmaceutical, biotechnology, food and beverage, aerospace and general lab applications. Saint Gobain Performance Plastics™ Silicone Tubing is peroxide-cured, offers excellent heat resistance and a low compression set.

  • Odorless, nontoxic, translucent tubing
  • Elastomer is FDA-compliant (21 CFR 177.2600) and meets USP Class VI requirements
  • Manufactured according to GMP
  • Sterilize by autoclave or gamma irradiation
Specifications

Specifications

Tubing
0.19 in.
Silicone
8 mm
7.6 m
1.6 mm
Translucent
230°c
Autoclavable
4.8 mm
0.31 in.
24.9 ft.
0.06 in.
-50°C
446°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.