Learn More
Trajan Scientific and Medical™ SilTite™ Ferrules
Ensures easy, leak-free installation for capillary columns without the use of any tools.
Brand:  Trajan Scientific and Medical™ 073332
 View more versions of this product
Product Code. 15288923
  • £55.90 / Pack of 10
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
SilTite Ferrules for 0.45 to 0.53 mm ID Column for Thermo Scientific
For Use with Thermo GC/MS Interface Connections 0.45 to 0.53 mm I.D. Columns
0.45 to 0.53 mm