Learn More
Thermo Scientific™ SilTite™ GC Capillary Column Connector
Use SilTite GC Capillary Column Connectors for fused silica capillary GC columns.
Brand:  Thermo Scientific™ 290MU500
 View more versions of this product
Product Code. 10301055
  • £95.12 / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use Thermo Scientific™ SilTite Capillary Column Connectors for fused silica capillary GC columns. They are glass lined for inertness and have a low thermal mass.
Join together two GC columns
Repair broken columns
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Capillary Column Connector
290 MU500
0.45 to 0.53 mm
Fused Silica Capillary Column
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.