missing translation for 'onlineSavingsMsg'
Learn More
Silver casting grain, 1.6-6.35mm (0.063-0.25in), 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11348498

Silver casting grain, 1.6-6.35mm (0.063-0.25in), 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11348498
100 g
Click to view available options
Quantity:
100 g
250 g
This item is not returnable. View return policy

Product Code. 11348498

Brand: Thermo Scientific Alfa Aesar 040511.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Questo articolo non è restituibile. Consulta la politica di reso

Jewelry, alloys, electrical contacts

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 7440-22-4
Molecular Formula Ag
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifica

Color Silver
Appearance Silver-colored grain
Assay Percent Range 99.9% (Metals basis)
Quantity 100 g
Molecular Weight (g/mol) 107.87
Odor Characteristic
Form Casting Grain
Chemical Name or Material Silver

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX03K002 Certificate TypeCertificate of Analysis Date21/03/2025 Catalog Number040511.22,040511.30
Lot NumberN06L018 Certificate TypeCertificate of Analysis Date10/02/2025 Catalog Number040511.A1,040511.22,040511.30
Lot NumberT19J034 Certificate TypeCertificate of Analysis Date25/07/2023 Catalog Number040511.22,040511.30
Lot NumberW01I017 Certificate TypeCertificate of Analysis Date16/09/2022 Catalog Number040511.22,040511.30,040520.04
Lot NumberR10I031 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog Number040511.22,040510.30,040510.09,040510.18

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
Silver casting grain, 1.6-6.35mm (0.063-0.25in), 99.9% (metals basis), Thermo Scientific Chemicals >

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato