missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver cyanide, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Silver cyanide is used in silver-plating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 320°C (decomposition) |
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Linear Formula | AgCN |
UN Number | UN1684 |
Merck Index | 14,8512 |
Solubility Information | Insoluble in water,dilute acids,and alcohol; Soluble in alkali cyanides and boiling concentrated HNO3 |
Formula Weight | 133.84 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction