missing translation for 'onlineSavingsMsg'
Learn More
Silver foil, 0.025mm (0.001in) thick, annealed, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11396578

Silver foil, 0.025mm (0.001in) thick, annealed, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11396578
150 x 150 mm
Click to view available options
Quantity:
150 x 150 mm
150 x 600 mm
This item is not returnable. View return policy

Product Code. 11396578

Brand: Thermo Scientific Alfa Aesar 012190.GT

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx. 5.9g/150 x 150mm; Temper: soft

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Color Silver
Appearance Silver-colored foil
Assay Percent Range 99.95% (Metals basis)
Quantity 150 x 150 mm
Odor Characteristic
Form Foil
Chemical Name or Material Silver, Annealed

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU13K012 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number012190.GT,012190.1!,012190.GV
Lot NumberT27G025 Certificate TypeCertificate of Analysis Date23/07/2024 Catalog Number012190.GT,012190.1!,012190.GV
Lot NumberX06J029 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number012190.GT,012190.1!,012190.GV
Lot NumberS22J011 Certificate TypeCertificate of Analysis Date28/06/2023 Catalog Number012190,012190.GT,012190.GV
Lot NumberR19I045 Certificate TypeCertificate of Analysis Date30/05/2022 Catalog Number012190.GT,012190.GV

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver foil, 0.025mm (0.001in) thick, annealed, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.