Learn More
Silver(I) oxide, 99+%, Thermo Scientific Chemicals
£56.80 - £676.00
Chemical Identifiers
CAS | 20667-12-3 |
---|---|
Molecular Formula | Ag2O |
Molecular Weight (g/mol) | 231.74 |
MDL Number | MFCD00003404 |
InChI Key | KHJDQHIZCZTCAE-UHFFFAOYSA-N |
IUPAC Name | (argentiooxy)silver |
SMILES | [Ag]O[Ag] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10450291
|
Thermo Scientific Acros
205800100 |
10 g | Glass bottle |
£56.80
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10751481
|
Thermo Scientific Acros
205800500 |
50 g | Glass bottle |
£182.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10608704
|
Thermo Scientific Acros
205802500 |
250 g | Glass bottle |
£676.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
20667-12-3 | |
231.74 | |
KHJDQHIZCZTCAE-UHFFFAOYSA-N | |
[Ag]O[Ag] |
Ag2O | |
MFCD00003404 | |
(argentiooxy)silver |
Specifications
20667-12-3 | |
Powder | |
Ag2O | |
MFCD00003404 | |
15, 8660 | |
KHJDQHIZCZTCAE-UHFFFAOYSA-N | |
(argentiooxy)silver | |
231.74 | |
Silver(I) oxide |
Black | |
99+% | |
Ag2O | |
01,1011; 02,368; 03,252; 04,430; 05,583; 06,515; 07,321; 08,442; 10,350; 11,468 | |
Solubility in water: slightly soluble. Other solubilities: freely soluble in dil.hno3 and ammonia,practically insoluble in alcohol | |
[Ag]O[Ag] | |
231.74 | |
≥99% |
Safety and Handling
GHS H Statement
Causes serious eye damage.
May cause fire or explosion; strong oxidizer.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 243-957-1
- Silver(I) oxide
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 1
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H271-May cause fire or explosion; strong oxidiser.
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
RUO – Research Use Only