missing translation for 'onlineSavingsMsg'
Learn More

Silver iodide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11391239
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Silver iodide, AgI, CAS Number-7783-96-2, colloidal silver iodide, silver iodide, ccris 8490, neosilvol, silver 1+ iodide, iodanylsilver, neosiluol, silver monoiodide, silver iodide metals basis, 100g, 99.9% (metals basis), 5.67 g/mL, Hazard Class: 9; Packaging Group: III, 232-038-0, 234.77

Used in medicine for its caustic, astringent and antiseptic effects. Used in photography, and in the estimation of silver. Through heterogeneous nucleation, it is used in cloud seeding, relying on its beta-AgI crystal structure being similar to that of Ice.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-96-2
Molecular Formula AgI
Molecular Weight (g/mol) 234.773
MDL Number MFCD00003412
InChI Key MSFPLIAKTHOCQP-UHFFFAOYSA-M
Synonym silver iodide, neosiluol, neosilvol, silver monoiodide, silver 1+ iodide, colloidal silver iodide, iodanylsilver, silver iodide metals basis, ccris 8490
PubChem CID 24563
IUPAC Name iodosilver
SMILES [Ag]I

Specifications

Melting Point 558°C
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Insoluble in water,acids and ammonium carbonate. Freely soluble in alkali cyanides and iodides.
Formula Weight 234.77
Sensitivity Light sensitive
Density 5.67 g/mL
Chemical Name or Material Silver iodide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'