Learn More
Silver nitrate, 0.1 N standard solution, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 124250010
63.75 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solutionSpecifications
Silver nitrate | |
7761-88-8, 7732-18-5 | |
Liquid | |
AgNO3 | |
MFCD00003414 | |
15, 8657 | |
Solubility in water: miscible. | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
CHEBI:32130 | |
Pure | |
HDPE Bottle | |
1.0150g/mL |
0.1 N Standard Solution | |
Colorless | |
1 L | |
AgNO3 | |
01,1008; 02,366; 03,252; 04,429; 05,582; 07,321; 09,411; 10,350; 11,268; 12,257; 14,350; 15,39 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver(1+) nitrate | |
24470 | |
169.87 | |
0.0998 to 0.1002N (20°C) | |
1.015 |
Safety and Handling
- Silver nitrate, 0.1N standard solution
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P390-Absorb spillage to prevent material damage.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only