missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, 0.1M, Reagecon™
Click to view available options
Quantity:
1 L
Specifications
Specifications
Quantity | 1 L |
Safety and Handling
Product Identifier
- SILVER NITRATE 0.1N (0.1M)
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P234-Keep only in original container.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P406-Store in corrosive resistant/ container with a resistant inner liner.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : silver nitrate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'