missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, 99.8+%, Extra Pure, SLR, Fisher Chemical™

Product Code. 11428822 Shop All Fisher Chemical Products
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
Unit Size:
100g
250g
25g
500g
This item is not returnable. View return policy

Product Code. 11428822

Brand: Fisher Chemical S/1240/50

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Boiling Point 444°C
Melting Point 212°C
Color White
pH 5.4 to 6.4
Physical Form Solid
Quantity 250 g
IUPAC Name silver;nitrate
Formula Weight 169.87g/mol
Packaging Glass Jar
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Silver nitrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.