missing translation for 'onlineSavingsMsg'
Learn More
Silver nitrate, 99.85%, for analysis, Thermo Scientific Chemicals Product Code.: 10449900

Silver nitrate, 99.85%, for analysis, Thermo Scientific Chemicals

Product Code. 10449900
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10449900

Brand: Thermo Scientific Acros 197680250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Assay Percent Range 99.85%
Linear Formula AgNO3
Merck Index 15, 8657
IUPAC Name silver(1+) nitrate
Formula Weight 169.87
Percent Purity ≥99.80%
Grade Analytical
Loss on Drying 0.1% (1g, 70°C, 2 hrs) max.
Packaging Glass bottle
Decomposition Information >444°C
Chemical Name or Material Silver nitrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468376 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number197680025,197681000,197680050,197680250,197685000
Lot NumberA0469077 Certificate TypeCertificate of Analysis Date10/01/2025 Catalog Number197680025,197681000,197680050,197680250,197685000
Lot NumberA0467063 Certificate TypeCertificate of Analysis Date23/10/2024 Catalog Number197680025,197681000,197680050,197680250,197685000
Lot NumberA0432340 Certificate TypeCertificate of Analysis Date15/01/2024 Catalog Number197680025,197681000,197680050,197680250,197685000
Lot NumberB0149836A Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number197680025,197681000,197680050,197680250,197685000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Silver nitrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver nitrate, 99.85%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.