missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver nitrate, 99.85%, for analysis, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
Specifications
Specifications
Boiling Point | 444.0°C |
Melting Point | 212.0°C |
Color | White |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 2.5 kg |
Assay Percent Range | 99.85% |
Linear Formula | AgNO3 |
Fieser | 01,1008; 02,366; 03,252; 04,429; 05,582; 07,321; 09,411; 10,350; 11,268; 12,257; 14,350; 15,39 |
Merck Index | 15, 8657 |
Solubility Information | Solubility in water: 219 g/100mL (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone and ether |
Show More |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Oxidising solid Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H360D-May damage the unborn child.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'