missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, Certified AR for Analysis, Fisher Chemical™

AgNO3, CAS Number-7761-88-8, silbernitrat, silver i nitrate, silver mononitrate, nitrate d'argent, silver nitrate, nitric acid silver i salt, silvernitrate, argenti nitras, argerol, lunar caustic, 500g, 444 deg.C, CHEBI:32130, White, 169.87g/mol, SQGYOTSLMSWVJD-UHFFFAOYSA-N, silver;nitrate

£271.00 - £3280.00

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
SMILES [Ag+].[O-][N+]([O-])=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10060340
View Documents Promotion Details
Fisher Chemical
S/1280/46
25 g
£271.00
25g
Please to purchase this item. Need a web account? Register with us today!
10764381
View Documents Promotion Details
Fisher Chemical
S/1280/48
100 g
£785.00
100g
Please to purchase this item. Need a web account? Register with us today!
10224350
View Documents Promotion Details
Fisher Chemical
S/1280/50
250 g
£1490.00
250g
Please to purchase this item. Need a web account? Register with us today!
10355860
View Documents Promotion Details
Fisher Chemical
S/1280/53
500 g
£3280.00
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AgNO3, CAS Number-7761-88-8, silbernitrat, silver i nitrate, silver mononitrate, nitrate d'argent, silver nitrate, nitric acid silver i salt, silvernitrate, argenti nitras, argerol, lunar caustic, 500g, 444 deg.C, CHEBI:32130, White, 169.87g/mol, SQGYOTSLMSWVJD-UHFFFAOYSA-N, silver;nitrate

Specifications

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
[Ag+].[O-][N+]([O-])=O
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130

Specifications

444°C
7761-88-8
5.4 to 6.4
AgNO3
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
[Ag+].[O-][N+]([O-])=O
169.87
CHEBI:32130
Glass Jar
212°C
White
Solid
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver;nitrate
24470
169.87g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Silver nitrate

Signal Word
  • Danger

Hazard Category
  • Oxidising solid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications