Learn More
Silver nitrate, Premion™, 99.995% (metals basis), Ag 63% min, Thermo Scientific Chemicals
£33.20 - £275.00
Chemical Identifiers
CAS | 7761-88-8 |
---|---|
Molecular Formula | AgNO3 |
Molecular Weight (g/mol) | 169.87 |
MDL Number | MFCD00003414 |
InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
Synonym | silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
PubChem CID | 24470 |
ChEBI | CHEBI:32130 |
IUPAC Name | silver(1+) nitrate |
SMILES | [Ag+].[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11394738
|
Thermo Scientific Alfa Aesar
043087.06 |
5 g |
£33.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304748
|
Thermo Scientific Alfa Aesar
043087.14 |
25 g |
£92.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11384738
|
Thermo Scientific Alfa Aesar
043087.22 |
100 g |
£275.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Silver nitrate is employed in Silver plating, photography, manufacturing of other silver compounds, mirrors, coloring porcelain, and staining in histology. It finds use as a stain in scanning electron microscopy, and a key ingredient in several silver-based antiseptic and antibacterial pharmaceutical compositions. Silver(I) nitrate can be used to separate mixtures of alkenes by selective absorption, and for deprotection and oxidation reactions in organic synthesis. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7761-88-8 | |
169.87 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
24470 | |
silver(1+) nitrate |
AgNO3 | |
MFCD00003414 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
CHEBI:32130 | |
[Ag+].[O-][N+]([O-])=O |
Specifications
212°C | |
White | |
AgNO3 | |
UN1479 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver(1+) nitrate | |
24470 | |
169.87 | |
Premion™ | |
Odorless | |
4.352 g/mL | |
Silver nitrate |
7761-88-8 | |
Crystalline | |
MFCD00003414 | |
14,8518 | |
Freely soluble in water,alcohol and aqueous ammonia. Slightly soluble in ether. | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
CHEBI:32130 | |
99.995% | |
Ag 63% min | |
Light sensitive | |
(metals basis) |
Safety and Handling
GHS H Statement
H272-H314-H290
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
May be corrosive to metals.
P220-P221-P234-P260-P262-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H272-H290-H314-H335-H373
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II
missing translation for 'einecsNumber' : 231-853-9
missing translation for 'rtecsNumber' : VW4725000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only