missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, puriss. p.a., Honeywell Fluka™

Product Code. 15687340 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
This item is not returnable. View return policy

Product Code. 15687340

Brand: Honeywell-Fluka 85228100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a.,≥99.5% (AT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Quantity 100 g
Linear Formula AgNO3
UN Number UN1493
Formula Weight 169.87g/mol
Percent Purity ≥99.5% (AT)
Grade puriss. p.a.
Density Vapor Density: 5.8 (vs air)
Chemical Name or Material Silver nitrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.