missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical

Product Code. 10366330 Shop All Fisher Chemical Products
Click to view available options
Quantity:
6 Ampules
Unit Size:
6 ampoules
This item is not returnable. View return policy

Product Code. 10366330

Brand: Fisher Chemical J/7330C/90

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Color Colorless
CAS 7761-88-8
Molecular Formula AgNO3
MDL Number MFCD00003414
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES [Ag+].[O-][N+]([O-])=O
IUPAC Name silver(1+) nitrate
Molecular Weight (g/mol) 169.87
PubChem CID 24470
ChEBI CHEBI:32130
Packaging Ampoule
pH 5.4 to 6.4
Quantity 6 Ampules
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.