missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver nitrite, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 140.0°C |
Color | Yellow to Green |
Physical Form | Powder and/or Agglomerates |
Quantity | 10 g |
Assay Percent Range | 98.5% min. (Argentometry) |
Linear Formula | AgNO2 |
Fieser | 01,1011; 06,515; 11,467 |
Merck Index | 15, 8658 |
Solubility Information | Solubility in water: 3.3g/L (20°C). Other solubilities: decomposes in diluted acids, insoluble in alcohol |
Formula Weight | 153.88 |
Show More |
Safety and Handling
Product Identifier
- Silver nitrite
- Danger
- Oxidising solid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'