missing translation for 'onlineSavingsMsg'
Learn More
Silver perchlorate, anhydrous, Thermo Scientific Chemicals Product Code.: 11369857

Silver perchlorate, anhydrous, Thermo Scientific Chemicals

Product Code. 11369857
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11369857

Brand: Thermo Scientific Alfa Aesar 042209.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Light Sensitive & Hygroscopic

Silver perchlorate is used as a catalyst in organic chemistry. It is an effective reagent for replacing halides ligands with perchlorate. It is also used in the manufacturing of explosives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-93-9
Molecular Formula AgClO4
Molecular Weight (g/mol) 207.314
MDL Number MFCD00003400
InChI Key YDHABVNRCBNRNZ-UHFFFAOYSA-M
Synonym silver perchlorate, unii-sv4x1u113o, silver 1+ ion perchlorate, perchloric acid, silver 1+ salt, silver 1+ salt, ag.clo4, perchloric acid, silver 1+ salt 1:1, silver 1+ perchlorate ion, silver i perchloratehydrate, silver i perchlorate, anhydrousShow MoreShow Less
PubChem CID 24562
IUPAC Name silver;perchlorate
SMILES [O-]Cl(=O)(=O)=O.[Ag+]

Specifications

Melting Point 485°C (decomposition)
Color White
Physical Form Crystalline
Quantity 25 g
UN Number UN1481
Merck Index 14,8523
Solubility Information Soluble in water and many organic solvents (benzene,toluene,pyridine)Soluble in water,benzene and toluene.
Formula Weight 207.32
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Density 2.806
Chemical Name or Material Silver perchlorate, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM10L046 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog Number042209.22,042209.06,042209.14
Lot NumberX04I062 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number042209.22,042209.06,042213.14,042209.14,042213.22
Lot NumberZ23H056 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number042209.22,042209.06,042213.14,042209.14,042213.22
Lot NumberJ11P08 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042209.22,042209.06,042213.14,042209.14,042213.22
Lot NumberF07Q17 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042209.22,042209.06,042213.14,042209.14,042213.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver perchlorate, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.