Learn More
Silver powder, -100 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
£139.00 - £430.00
Chemical Identifiers
CAS | 7440-22-4 |
---|---|
Molecular Formula | Ag |
Molecular Weight (g/mol) | 107.87 |
MDL Number | MFCD00003397 |
InChI Key | BQCADISMDOOEFD-UHFFFAOYSA-N |
Synonym | argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber |
PubChem CID | 23954 |
ChEBI | CHEBI:9141 |
IUPAC Name | silver |
SMILES | [Ag] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11336627
|
Thermo Scientific Alfa Aesar
000690.14 |
25 g |
£139.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11326627
|
Thermo Scientific Alfa Aesar
000690.22 |
100 g |
£430.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
7440-22-4 | |
107.87 | |
BQCADISMDOOEFD-UHFFFAOYSA-N | |
23954 | |
silver |
Ag | |
MFCD00003397 | |
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
CHEBI:9141 | |
[Ag] |
Especificaciones
Silver | |
Silver-colored powder | |
Ag | |
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
[Ag] | |
107.87 | |
CHEBI:9141 | |
Characteristic | |
Silver |
7440-22-4 | |
99.95% (Metals basis) | |
MFCD00003397 | |
BQCADISMDOOEFD-UHFFFAOYSA-N | |
silver | |
23954 | |
-100 Mesh | |
Powder |
Seguridad y manipulación
H400-H410; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III
missing translation for 'einecsNumber' : 231-131-3
missing translation for 'rtecsNumber' : VW3500000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Silver powder
Palabra de advertencia
- Warning
Clasificación
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Indicaciones de peligro
- H410-Very toxic to aquatic life with long lasting effects.
Consejos de prudencia
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501-Dispose of contents/container to .
RUO – Research Use Only