missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Silver Protein for Histology, Strong

Product Code. p-7070065
Click to view available options
Quantity:
5g
25g
110g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11375018

Brand: Thermo Scientific Chemicals 043970.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9008-42-8

Silver protein is used in electron microscopy with periodic acid and thiocarbohydrazide or thiosemicarbohydrazide as a positive stain for carbohydrates such as glycogen. It can also be used for light microscopy to stain nerve tissue. It is also used in histological stain for microscopic analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9008-42-8
Synonym Protargol

Specifications

Color Pale yellow to brown
Form Powder
Format Powder
For Use With (Application) Histological stain for microscopic analysis
Product Type Silver Protein
Quantity 5g
Sensitivity Hygroscopic; Light sensitive

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.