missing translation for 'onlineSavingsMsg'
Learn More

Silver rod, 12.5mm (0.492in) dia, Premion™, 99.9985% (metals basis), Thermo Scientific Chemicals

Product Code. 11391129
Click to view available options
Quantity:
2 cm
10 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 2 cm
Percent Purity 99.9985%
Grade Premion™
Odor Characteristic
Weight ≈12.87g/cm
Form Rod
Assay (metals basis)
Chemical Name or Material Silver rod, 12.5mm (0.492 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'