missing translation for 'onlineSavingsMsg'
Learn More
Silver shot, 1-5mm (0.04-0.2in), 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11396419

Silver shot, 1-5mm (0.04-0.2in), 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11396419
10 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11396419

Brand: Thermo Scientific Alfa Aesar 011436.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Color Silver
Appearance Silver-colored shot
Assay Percent Range 99.9% (Metals basis)
Quantity 10 g
Odor Characteristic
Form Shot
Chemical Name or Material Silver

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY04K043 Certificate TypeCertificate of Analysis Date04/12/2024 Catalog Number011436.09,011436.30,011436.18
Lot NumberX07K003 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number011436.09,011436.30,011436.18
Lot NumberT24J057 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog Number011436.09,011436.30,011436.18
Lot NumberN24J010 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number011436.09
Lot NumberN21I055 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011555.30,011436.09,011436.30,011436.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver shot, 1-5mm (0.04-0.2in), 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.