Learn More
Silver, solder alloy, 1.6mm (0.06in) dia, Thermo Scientific Chemicals
For use on copper, stainless, and most other metals
£29.00 - £128.00
Specifications
Chemical Name or Material | Silver, solder Alloy |
---|---|
Name Note | 1.6mm (0.06 in.) dia. |
Avg. Mol. Wt. or Mol. Wt. Range | Ag:Sn; 3.5:96.5 wt% |
MDL Number | MFCD00801108 |
Solubility Information | Insoluble in water. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11349447
|
Thermo Scientific Alfa Aesar
041032.18 |
50 g |
£29.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11339447
|
Thermo Scientific Alfa Aesar
041032.30 |
250 g |
£74.80
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11329447
|
Thermo Scientific Alfa Aesar
041032.36 |
500 g |
£128.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Silver, solder alloy is used on copper, stainless, and most other metals. It is used for electronic assemblies, die attachments, thick films. Soldering of jewellery due to the alloys ability to maintain high levels of brightness.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver, solder Alloy | |
Ag:Sn; 3.5:96.5 wt% | |
Insoluble in water. |
1.6mm (0.06 in.) dia. | |
MFCD00801108 |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only