missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Standard Metal Solution 1000ppm, for ICP, Solution HNO3 1M, Fisher Chemical
Click to view available options
Quantity:
500 mL
Packaging:
HDPE plastic bottle
Specifications
Specifications
CAS | 7761-88-8 |
Packaging | HDPE plastic bottle |
Quantity | 500 mL |
Color | Colorless |
Molecular Formula | AgNO3 |
MDL Number | MFCD00003414 |
InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
SMILES | [Ag+].[O-][N+]([O-])=O |
IUPAC Name | silver;nitrate |
Molecular Weight (g/mol) | 169.87 |
Show More |
Safety and Handling
Product Identifier
- Silver (I) single element solution 1,000
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
- P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'