Learn More
Silver standard solution, for AAS, 1mg/mL Ag in 0.5N HNO3, Thermo Scientific Chemicals
Marke: Thermo Scientific Acros 196301000
Code : 29
Weitere Details : CAS Number : 7761-88-8 Weight : 0.10200kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopySpezifikation
Silver standard solution | |
1 mg/ml Ag in 0.5N HNO3 | |
Plastic bottle | |
Colorless | |
MFCD00003414 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver(1+) nitrate | |
24470 | |
107.87 | |
1.0200g/mL | |
101.0°C | |
Soluble in water |
997 to 1003 μg/mL of Ag (1+) in 0.5N HNO3 (20°C) | |
7761-88-8,7697-37-2,7732-18-5 | |
100 mL | |
AgNO3 | |
15, 8657 | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
CHEBI:32130 | |
AAS | |
-3.0°C | |
1.02 | |
Liquid |
RUO – Research Use Only