missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver standard solution, for AAS, 1mg/mL Ag in 0.5N HNO3, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 196305000
43.65 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopySpecifications
Silver standard solution | |
1 mg/ml Ag in 0.5N HNO3 | |
Plastic bottle | |
Colorless | |
MFCD00003414 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver(1+) nitrate | |
24470 | |
107.87 | |
1.0200g/mL | |
101.0°C | |
Soluble in water |
997 to 1003 μg/mL of Ag (1+) in 0.5N HNO3 (20°C) | |
7761-88-8, 7697-37-2, 7732-18-5 | |
500 mL | |
AgNO3 | |
15, 8657 | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
CHEBI:32130 | |
AAS | |
-3.0°C | |
1.02 | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Silver atomic absorption standard solution
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : Nitric acid, Silvernitrate
RUO – Research Use Only