Learn More
Silver sulfadiazine, 98%, Thermo Scientific Chemicals
£44.50 - £150.00
Chemical Identifiers
CAS | 22199-08-2 |
---|---|
Molecular Formula | C10H9AgN4O2S |
Molecular Weight (g/mol) | 357.14 |
InChI Key | UEJSSZHHYBHCEL-UHFFFAOYSA-N |
Synonym | silver sulfadiazine, sulfadiazine silver, flamazine, silver sulphadiazine, silvadene, sulfadiazine silver salt, dermazin, thermazene, silver sulfadiazinate, sulfadiazine, silver |
PubChem CID | 441244 |
ChEBI | CHEBI:9142 |
IUPAC Name | silver;(4-aminophenyl)sulfonyl-pyrimidin-2-ylazanide |
SMILES | C1=CN=C(N=C1)[N-]S(=O)(=O)C2=CC=C(C=C2)N.[Ag+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15569493
|
Thermo Scientific Acros
459650050 |
5 g |
£44.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15579493
|
Thermo Scientific Acros
459650250 |
25 g |
£150.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
22199-08-2 | |
357.14 | |
silver sulfadiazine, sulfadiazine silver, flamazine, silver sulphadiazine, silvadene, sulfadiazine silver salt, dermazin, thermazene, silver sulfadiazinate, sulfadiazine, silver | |
CHEBI:9142 | |
C1=CN=C(N=C1)[N-]S(=O)(=O)C2=CC=C(C=C2)N.[Ag+] |
C10H9AgN4O2S | |
UEJSSZHHYBHCEL-UHFFFAOYSA-N | |
441244 | |
silver;(4-aminophenyl)sulfonyl-pyrimidin-2-ylazanide |
Specifications
285.0°C | |
97.5% min. (HPLC) | |
silver sulfadiazine, sulfadiazine silver, flamazine, silver sulphadiazine, silvadene, sulfadiazine silver salt, dermazin, thermazene, silver sulfadiazinate, sulfadiazine, silver | |
C1=CN=C(N=C1)[N-]S(=O)(=O)C2=CC=C(C=C2)N.[Ag+] | |
357.14 | |
CHEBI:9142 | |
98% | |
0.5% max. |
22199-08-2 | |
C10H9AgN4O2S | |
UEJSSZHHYBHCEL-UHFFFAOYSA-N | |
silver;(4-aminophenyl)sulfonyl-pyrimidin-2-ylazanide | |
441244 | |
357.14 | |
Authentic | |
Silver sulfadiazine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 244-834-5
- Silver sulfadiazine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only