missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver sulfate, 99%, extra pure, Thermo Scientific Chemicals
45.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 1085.0°C |
Melting Point | 652.0°C |
Color | White |
Physical Form | Fine Crystalline Powder |
Quantity | 10 g |
Assay Percent Range | 98.5% min. (Argentometry) |
Linear Formula | Ag2SO4 |
Fieser | 01,1015; 03,254; 04,435 |
Merck Index | 15, 8668 |
Solubility Information | Solubility in water: 7.4g/L (20°C). Other solubilities: soluble in nh4oh, nitric acid and sulfuric acid, insoluble in alcohol |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'