missing translation for 'onlineSavingsMsg'
Learn More

Silver sulfate, 99%, extra pure, Thermo Scientific Chemicals

Product Code. 10783884
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-26-5
Molecular Formula Ag2H2O4S
Molecular Weight (g/mol) 313.81
MDL Number MFCD00003407
InChI Key YPNVIBVEFVRZPJ-UHFFFAOYSA-N
Synonym silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
PubChem CID 159865
IUPAC Name disilver(1+) sulfuric acid
SMILES [Ag+].[Ag+].OS(O)(=O)=O

Specifications

Boiling Point 1085.0°C
Melting Point 652.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 500 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula Ag2SO4
Fieser 01,1015; 03,254; 04,435
Merck Index 15, 8668
Solubility Information Solubility in water: 7.4g/L (20°C). Other solubilities: soluble in nh4oh,nitric acid and sulfuric acid,insoluble in alcohol
Formula Weight 311.79
Percent Purity 99%
Grade Extra Pure
Packaging Glass bottle
Density 1.84g/mL
Chemical Name or Material Silver sulfate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.