Learn More
Silver sulfate, ACS reagent, Thermo Scientific Chemicals
£39.00 - £970.00
Chemical Identifiers
CAS | 10294-26-5 |
---|---|
Molecular Formula | Ag2H2O4S |
Molecular Weight (g/mol) | 313.81 |
InChI Key | YPNVIBVEFVRZPJ-UHFFFAOYSA-N |
Synonym | silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt |
PubChem CID | 159865 |
IUPAC Name | disilver(1+) sulfuric acid |
SMILES | [Ag+].[Ag+].OS(O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13226309
|
Thermo Scientific Acros
194070100 |
10 g |
£39.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13266309
|
Thermo Scientific Acros
194071000 |
100 g |
£222.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13256309
|
Thermo Scientific Acros
194075000 |
500 g |
£970.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10294-26-5 | |
313.81 | |
silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt | |
disilver(1+) sulfuric acid |
Ag2H2O4S | |
YPNVIBVEFVRZPJ-UHFFFAOYSA-N | |
159865 | |
[Ag+].[Ag+].OS(O)(=O)=O |
Specifications
1085.0°C | |
10294-26-5 | |
Ag2SO4 | |
15,8668 | |
Solubility in water: 7.4g/L (20°C). Other solubilities: soluble in nh4oh, nitric acid and sulfuric acid, insoluble in alcohol | |
[Ag+].[Ag+].OS(O)(=O)=O | |
313.81 | |
311.79 | |
Glass bottle |
652.0°C | |
Ag2H2O4S | |
01,1015; 03,254; 04,435 | |
silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt | |
YPNVIBVEFVRZPJ-UHFFFAOYSA-N | |
disilver(1+) sulfuric acid | |
159865 | |
ACS Reagent | |
Silver sulfate |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 233-653-7
RUO – Research Use Only