missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Sulfate, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Amber glass bottle
Glass Jar
Specifications
Specifications
Boiling Point | 1085°C |
Melting Point | 652°C |
Color | White |
pH | 5 to 6 |
Physical Form | Solid |
Quantity | 25 g |
Formula Weight | 311.79g/mol |
Packaging | Glass Jar |
Safety and Handling
Product Identifier
- Silver sulfate
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'