missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Sulfate, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Amber glass bottle
Glass Jar
Specifications
Boiling Point | 1085°C |
Melting Point | 652°C |
Color | White |
pH | 5 to 6 |
Physical Form | Solid |
Quantity | 100 g |
Formula Weight | 311.79g/mol |
Packaging | Glass Jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'