Learn More
Silver sulfate, Premion™, 99.99% (metals basis), Ag 68.9% min, Thermo Scientific Chemicals
£93.80 - £334.00
Chemical Identifiers
CAS | 10294-26-5 |
---|---|
Molecular Formula | Ag2H2O4S |
Molecular Weight (g/mol) | 313.81 |
MDL Number | MFCD00003407 |
InChI Key | YPNVIBVEFVRZPJ-UHFFFAOYSA-N |
Synonym | silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt |
PubChem CID | 159865 |
IUPAC Name | disilver(1+) sulfuric acid |
SMILES | [Ag+].[Ag+].OS(O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11324168
|
Thermo Scientific Alfa Aesar
041443.14 |
25 g |
£93.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11334168
|
Thermo Scientific Alfa Aesar
041443.22 |
100 g |
£334.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Silver(I) sulfate is employed in silver plating, and in electrolysis. It is a useful non-staining substitute to silver nitrate. Silver sulfate/iodine is an excellent iodination reagent for alkyl, alkoxybenzenes, aromatic amines and uridines. It is used in batteries; its pharmaceutical composition is used as an antimicrobial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10294-26-5 | |
313.81 | |
YPNVIBVEFVRZPJ-UHFFFAOYSA-N | |
159865 | |
[Ag+].[Ag+].OS(O)(=O)=O |
Ag2H2O4S | |
MFCD00003407 | |
silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt | |
disilver(1+) sulfuric acid |
Specifications
1085°C (decomposition) | |
10294-26-5 | |
Powder | |
Ag2SO4 | |
UN3077 | |
silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt | |
YPNVIBVEFVRZPJ-UHFFFAOYSA-N | |
disilver(1+) sulfuric acid | |
159865 | |
99.99% | |
Ag 68.9% min. | |
Light sensitive | |
(metals basis) |
657°C | |
White | |
Ag2H2O4S | |
MFCD00003407 | |
14,8529 | |
Partially soluble in water. Water solubility increases with temperature. Soluble in nitric acid,ammonia and conc. sulfuric acid. | |
[Ag+].[Ag+].OS(O)(=O)=O | |
313.81 | |
311.8 | |
Premion™ | |
Odorless | |
5.45 g/mL | |
Silver sulfate |
Safety and Handling
GHS H Statement
H318
Causes serious eye damage.
P280-P305+P351+P338-P310
H318
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 233-653-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only